Anti-GJB2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7853-03BEA 3290 DKK
USBIC7853-03B
Anti-GJB2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot:1-2?g/ml

Type: Primary
Antigen: GJB2 (gap junction protein, beta 2, 26kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR