Anti-Conalbumin, Chicken Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7851-90EA 5040 DKK
USBIC7851-90
Anti-Conalbumin, Chicken Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:10000-1:50000; Western Blot: 1:1000-1:5000

Type: Primary
Antigen: Conalbumin, Chicken
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR