Anti-COMT Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7851-70NEA 4320 DKK
USBIC7851-70N
Anti-COMT Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 5-7. 5 ?g/ml

Type: Primary
Antigen: COMT (catechol-O-methyltransferase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR