Anti-Complement Factor P Mouse Monoclonal Antibody [clone: 10B2571]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-95EA 4850 DKK
USBIC7850-95
Anti-Complement Factor P Mouse Monoclonal Antibody [clone: 10B2571]
Antistoffer
Type: Primary
Antigen: Complement Factor P
Clonality: Monoclonal
Clone: 10B2571
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR