Anti-Complement Factor H-Related Protein 5 Mouse Monoclonal Antibody [clone: 7H187]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-88EA 4320 DKK
USBIC7850-88
Anti-Complement Factor H-Related Protein 5 Mouse Monoclonal Antibody [clone: 7H187]
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: Complement Factor H-Related Protein 5
Clonality: Monoclonal
Clone: 7H187
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR