Anti-CFH Mouse monoclonal antibody [clone: 6F85]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-80EA 5480 DKK
USBIC7850-80
Anti-CFH Mouse monoclonal antibody [clone: 6F85]
Antistoffer
Recommended dilution: ELISA: 1:40000

Type: Primary
Antigen: CFH (complement factor H)
Clonality: Monoclonal
Clone: 6F85
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR