Anti-Complement Factor D Sheep Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-71AEA 5420 DKK
USBIC7850-71A
Anti-Complement Factor D Sheep Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: Western Blot: 0.1 ?g/ml

Type: Primary
Antigen: Complement Factor D
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR