Anti-Complement Factor Bb Mouse Monoclonal Antibody [clone: 10B2568]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-68EA 5520 DKK
USBIC7850-68
Anti-Complement Factor Bb Mouse Monoclonal Antibody [clone: 10B2568]
Antistoffer
Type: Primary
Antigen: Complement Factor Bb
Clonality: Monoclonal
Clone: 10B2568
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR