Anti-CFB Mouse monoclonal antibody [clone: 10K168]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-60JEA 3590 DKK
USBIC7850-60J
Anti-CFB Mouse monoclonal antibody [clone: 10K168]
Antistoffer
Recommended dilution: ELISA: 1 ?g/ml; ELISA: 5 ?g/ml

Type: Primary
Antigen: CFB (complement factor B)
Clonality: Monoclonal
Clone: 10K168
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR