Anti-CFB Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-60HEA 4380 DKK
USBIC7850-60H
Anti-CFB Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: CFB (complement factor B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR