Anti-Complement Factor B, Ba Fragment Mouse Monoclonal Antibody [clone: 10B2567]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-60DEA 4620 DKK
USBIC7850-60D
Anti-Complement Factor B, Ba Fragment Mouse Monoclonal Antibody [clone: 10B2567]
Antistoffer
Type: Primary
Antigen: Complement Factor B, Ba Fragment
Clonality: Monoclonal
Clone: 10B2567
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR