Anti-CFB Mouse monoclonal antibody [clone: 10K167]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-60BEA 3290 DKK
USBIC7850-60B
Anti-CFB Mouse monoclonal antibody [clone: 10K167]
Antistoffer
Type: Primary
Antigen: CFB (complement factor B)
Clonality: Monoclonal
Clone: 10K167
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR