Anti-Complement C9 Mouse Monoclonal Antibody [clone: X197]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-51EEA 6250 DKK
USBIC7850-51E
Anti-Complement C9 Mouse Monoclonal Antibody [clone: X197]
Antistoffer
Recommended dilution: Flow Cytometry: 1:10; Western Blot: 1:10; Immunochemistry: 1:10

Type: Primary
Antigen: Complement C9
Clonality: Monoclonal
Clone: X197
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR