Anti-C5AR1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11n36]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-24UEA 4500 DKK
USBIC7850-24U
Anti-C5AR1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11n36]
Antistoffer
Do not freeze!

Type: Primary
Antigen: C5AR1 (complement component 5a receptor 1)
Clonality: Monoclonal
Clone: 11n36
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR