Anti-Complement C5, Human Mouse Monoclonal Antibody [clone: 8J448]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-24MEA 4550 DKK
USBIC7850-24M
Anti-Complement C5, Human Mouse Monoclonal Antibody [clone: 8J448]
Antistoffer
Recommended dilution: Western Blot: 1:100-1:200

Type: Primary
Antigen: Complement C5, Human
Clonality: Monoclonal
Clone: 8J448
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR