Anti-Complement C5a des-Arg Mouse Monoclonal Antibody [clone: 6A534]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-24GEA 4410 DKK
USBIC7850-24G
Anti-Complement C5a des-Arg Mouse Monoclonal Antibody [clone: 6A534]
Antistoffer
Recommended dilution: Western Blot: 1:50

Type: Primary
Antigen: Complement C5a des-Arg
Clonality: Monoclonal
Clone: 6A534
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR