Anti-C5 Mouse monoclonal antibody [clone: 10B398]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-23EEA 4620 DKK
USBIC7850-23E
Anti-C5 Mouse monoclonal antibody [clone: 10B398]
Antistoffer
Type: Primary
Antigen: C5 (complement component 5)
Clonality: Monoclonal
Clone: 10B398
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR