Anti-Complement C5, Human Mouse Monoclonal Antibody [clone: 5F207]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-23EA 4290 DKK
USBIC7850-23
Anti-Complement C5, Human Mouse Monoclonal Antibody [clone: 5F207]
Antistoffer
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: Complement C5, Human
Clonality: Monoclonal
Clone: 5F207
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR