Anti-Complement Component 4 Binding Protein Sheep Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC7850-19FEA 4790 DKK
USBIC7850-19F
Anti-Complement Component 4 Binding Protein Sheep Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Complement Component 4 Binding Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human