Anti-Complement Component 4 Binding Protein Mouse Monoclonal Antibody [clone: 10B393]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-19D1EA 4780 DKK
USBIC7850-19D1
Anti-Complement Component 4 Binding Protein Mouse Monoclonal Antibody [clone: 10B393]
Antistoffer
Type: Primary
Antigen: Complement Component 4 Binding Protein
Clonality: Monoclonal
Clone: 10B393
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR