Anti-Complement Component 4 Binding Protein Sheep Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-19CEA 4610 DKK
USBIC7850-19C
Anti-Complement Component 4 Binding Protein Sheep Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:20-1:200

Type: Primary
Antigen: Complement Component 4 Binding Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR