Anti-Complement Component 4d Mouse Monoclonal Antibody [clone: 10B396]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-18HEA 7760 DKK
USBIC7850-18H
Anti-Complement Component 4d Mouse Monoclonal Antibody [clone: 10B396]
Antistoffer
Recommended dilution: Immunochemistry: 1:100-1:750; ELISA: 1:5000-1:20000; Immunofluorescence: 1:250-1:600

Type: Primary
Antigen: Complement Component 4d
Clonality: Monoclonal
Clone: 10B396
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR