Anti-Complement C4 Mouse Monoclonal Antibody [clone: 25.3E3]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-16EEA 3150 DKK
USBIC7850-16E
Anti-Complement C4 Mouse Monoclonal Antibody [clone: 25.3E3]
Antistoffer
Recommended dilution: ELISA: 1:4000

Type: Primary
Antigen: Complement C4
Clonality: Monoclonal
Clone: 25300
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR