Anti-Complement C4 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-15DEA 3290 DKK
USBIC7850-15D
Anti-Complement C4 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1-5 ?g/ml; Western Blot: 1:1000-1:5000

Type: Primary
Antigen: Complement C4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR