Anti-Complement C3, C3d Mouse Monoclonal Antibody [clone: 10B389]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-14YEA 5520 DKK
USBIC7850-14Y
Anti-Complement C3, C3d Mouse Monoclonal Antibody [clone: 10B389]
Antistoffer
Type: Primary
Antigen: Complement C3, C3d
Clonality: Monoclonal
Clone: 10B389
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR