Anti-Complement C3, C3c Goat Polyclonal Antibody (FITC (Fluorescein))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-14S7EA 3700 DKK
USBIC7850-14S7
Anti-Complement C3, C3c Goat Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: Complement C3, C3c
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Guinea pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR