Anti-Complement C3, C3c Mouse Monoclonal Antibody [clone: 10B386]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-14NEA 4620 DKK
USBIC7850-14N
Anti-Complement C3, C3c Mouse Monoclonal Antibody [clone: 10B386]
Antistoffer
Type: Primary
Antigen: Complement C3, C3c
Clonality: Monoclonal
Clone: 10B386
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR