Anti-Complement C3c Sheep Polyclonal Antibody (FITC (Fluorescein))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-14C1EA 3790 DKK
USBIC7850-14C1
Anti-Complement C3c Sheep Polyclonal Antibody (FITC (Fluorescein))
Recommended dilution: Immunofluorescence: 1:10-1:50; FLISA: 1:500-1:50000

Type: Primary
Antigen: Complement C3c
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR