Anti-C3AR1 Mouse monoclonal antibody [clone: 13A33]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-13T7EA 5810 DKK
USBIC7850-13T7
Anti-C3AR1 Mouse monoclonal antibody [clone: 13A33]
Antistoffer
Recommended dilution: Flow Cytometry: 1:50

Type: Primary
Antigen: C3AR1 (complement component 3a receptor 1)
Clonality: Monoclonal
Clone: 13A33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR