Anti-Complement C3a Receptor Mouse Monoclonal Antibody [clone: 10B380]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-13T1EA 3580 DKK
USBIC7850-13T1
Anti-Complement C3a Receptor Mouse Monoclonal Antibody [clone: 10B380]
Antistoffer
Recommended dilution: Flow Cytometry: 1:10-1:20;

Type: Primary
Antigen: Complement C3a Receptor
Clonality: Monoclonal
Clone: 10B380
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR