Anti-Complement C3a Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11C9]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-13LEA 5470 DKK
USBIC7850-13L
Anti-Complement C3a Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11C9]
Antistoffer
Do not freeze!

Type: Primary
Antigen: Complement C3a
Clonality: Monoclonal
Clone: 11C9
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR