Anti-Complement C3, C3a Mouse Monoclonal Antibody [clone: 10B377]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-13B1EA 6030 DKK
USBIC7850-13B1
Anti-Complement C3, C3a Mouse Monoclonal Antibody [clone: 10B377]
Antistoffer
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: Complement C3, C3a
Clonality: Monoclonal
Clone: 10B377
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Guinea pig, Human, Rabbit
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR