Anti-C3 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-12HEA 4650 DKK
USBIC7850-12H
Anti-C3 Goat polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000-1:5000; ELISA: 1:10000-1:50000

Type: Primary
Antigen: C3 (complement component 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR