Anti-C3 Mouse monoclonal antibody [clone: 6F67]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC7850-10KEA 5490 DKK
USBIC7850-10K
Anti-C3 Mouse monoclonal antibody [clone: 6F67]
Antistoffer
Recommended dilution: ELISA: 1:2000
Type: Primary
Antigen: C3 (complement component 3)
Clonality: Monoclonal
Clone: 6F67
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Rat