Anti-C3 Mouse monoclonal antibody [clone: 6F65]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-10HEA 3450 DKK
USBIC7850-10H
Anti-C3 Mouse monoclonal antibody [clone: 6F65]
Antistoffer
Type: Primary
Antigen: C3 (complement component 3)
Clonality: Monoclonal
Clone: 6F65
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR