Anti-Complement C2 Mouse Monoclonal Antibody [clone: 6F62]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-09BEA 5010 DKK
USBIC7850-09B
Anti-Complement C2 Mouse Monoclonal Antibody [clone: 6F62]
Antistoffer
Type: Primary
Antigen: Complement C2
Clonality: Monoclonal
Clone: 6F62
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR