Anti-Complement 1s Mouse Monoclonal Antibody [clone: 9G173]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-08FEA 4690 DKK
USBIC7850-08F
Anti-Complement 1s Mouse Monoclonal Antibody [clone: 9G173]
Antistoffer
Recommended dilution: Flow Cytometry: 1:10; Western Blot: 1:10; Immunochemistry: 1:10; Immunoprecipitation: 1:10

Type: Primary
Antigen: Complement 1s
Clonality: Monoclonal
Clone: 9G173
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR