Anti-Complement C1q, Human Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7850-06MEA 2900 DKK
USBIC7850-06M
Anti-Complement C1q, Human Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Complement C1q, Human
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR