Anti-COMP Chicken Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7849-09D-1EA 2440 DKK
USBIC7849-09D-1
Anti-COMP Chicken Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: COMP (cartilage oligomeric matrix protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR