Anti-COMMD1 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2565]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7847-50-FITCEA 0 DKK
USBIC7847-50-FITC
Anti-COMMD1 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2565]
Type: Primary
Antigen: COMMD1 (copper metabolism (Murr1) domain containing 1)
Clonality: Monoclonal
Clone: 10B2565
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR