Anti-Cre Recombinase Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7800-01EA 3500 DKK
USBIC7800-01
Anti-Cre Recombinase Rabbit polyclonal antibody
Antistoffer
Type: Primary
Antigen: Cre Recombinase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR