Anti-Colon Carcinoma Mouse Monoclonal Antibody [clone: CC-3]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7554-50BEA 3960 DKK
USBIC7554-50B
Anti-Colon Carcinoma Mouse Monoclonal Antibody [clone: CC-3]
Antistoffer
Recommended dilution: Immunochemistry: 1:10 Stains acetone or ethanol fixed tissues

Type: Primary
Antigen: Colon Carcinoma
Clonality: Monoclonal
Clone: CC-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR