Anti-Collapsin Response Mediator Protein 4 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10C8]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7517-08J-PEEA 0 DKK
USBIC7517-08J-PE
Anti-Collapsin Response Mediator Protein 4 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10C8]
Type: Primary
Antigen: Collapsin Response Mediator Protein 4
Clonality: Monoclonal
Clone: 10C8
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR