Anti-Collapsin Response Mediator Protein 4 Mouse monoclonal antibody Biotin [clone: 10C8]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7517-08J-BIOTEA 0 DKK
USBIC7517-08J-BIOT
Anti-Collapsin Response Mediator Protein 4 Mouse monoclonal antibody Biotin [clone: 10C8]
Type: Primary
Antigen: Collapsin Response Mediator Protein 4
Clonality: Monoclonal
Clone: 10C8
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR