Anti-CRMP1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7517-02EA 3340 DKK
USBIC7517-02
Anti-CRMP1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CRMP1 (collapsin response mediator protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR