Anti-Collagenase Rabbit Polyclonal Antibody (Biotin)
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC7511-06BEA 4540 DKK
USBIC7511-06B
Anti-Collagenase Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Collagenase
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu