Anti-COL13 Sheep Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-94AEA 4780 DKK
USBIC7510-94A
Anti-COL13 Sheep Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: COL13 (collagen, type XIII)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR