Anti-COL18A1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-66MEA 3700 DKK
USBIC7510-66M
Anti-COL18A1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: COL18A1 (collagen, type XVIII, alpha 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR