Anti-COL6 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-60LEA 4320 DKK
USBIC7510-60L
Anti-COL6 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: COL6 (collagen, type VI)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR