Anti-COL5 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2290]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-56N-APEA 0 DKK
USBIC7510-56N-AP
Anti-COL5 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2290]
Type: Primary
Antigen: COL5 (collagen, type V)
Clonality: Monoclonal
Clone: 10B2290
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR